1-Benzyl-1H-benzo[d][1,2,3]triazole 95%

Supplier: Ambeed
Warning

A144347-250MG A144347-1G A144347-5G
77010-480EA 17 USD
77010-480 77889-959 77889-960
1-Benzyl-1H-benzo[d][1,2,3]triazole 95%
1-Benzyl-1H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₁₃H₁₁N₃
MDL Number: MFCD00227101
CAS Number: 4706-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR