2,3,6-Trichloro-5-nitropyridine 95%

Supplier: Ambeed
Warning

A144285-100MG A144285-1G A144285-250MG A144285-5G
77027-776EA 20.16 USD
77027-776 77027-778 77027-780 77027-782
2,3,6-Trichloro-5-nitropyridine 95%
2,3,6-Trichloro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅HCl₃N₂O₂
MW: 227.43 g/mol
MDL Number: MFCD11227172
CAS Number: 55304-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR