4,4'-Diamino-2,2'-bipyridyl 97%

Supplier: Ambeed
Warning

Synonyms: [2,2'-Bipyridine]-4,4'-diamine, 4,4'-Diamino-2,2'-bipyridine

A144149-100MG A144149-1G A144149-250MG A144149-5G
76984-352EA 18.04 USD
76984-352 76984-354 76984-356 76984-358
4,4'-Diamino-2,2'-bipyridyl 97%
4,4'-Diamino-2,2'-bipyridyl

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₄
MW: 186.22 g/mol
MDL Number: MFCD08704188
CAS Number: 18511-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR