2-(5-Chloro-3-fluoropyridin-2-yl)acetonitrile 98%

Supplier: Ambeed
Warning

A144133-100MG A144133-250MG
76984-348EA 13 USD
76984-348 76984-350
2-(5-Chloro-3-fluoropyridin-2-yl)acetonitrile 98%
2-(5-Chloro-3-fluoropyridin-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₇H₄ClFN₂
MDL Number: MFCD16608835
CAS Number: 1227572-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR