4-Amino-2-bromopyrimidine 95%

Supplier: Ambeed
Warning

A144113-10G A144113-1G A144113-250MG A144113-25G A144113-5G
77027-760EA 449.83 USD
77027-760 77027-762 77027-764 77027-766 77027-768
4-Amino-2-bromopyrimidine 95%
4-Amino-2-bromopyrimidine

Precaución: for Research Use Only

Formula: C₄H₄BrN₃
MW: 174.0000 g/mol
MDL Number: MFCD11109798
CAS Number: 1160994-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR