Methyl 4-bromo-5-fluoro-2-nitrobenzoate 98%

Supplier: Ambeed
Warning

A144083-100G A144083-100MG A144083-10G A144083-1G A144083-250MG A144083-25G A144083-5G
76984-332EA 392.54 USD
76984-332 76984-334 76984-336 76984-338 76984-340 76984-342 76984-344
Methyl 4-bromo-5-fluoro-2-nitrobenzoate 98%
Methyl 4-bromo-5-fluoro-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrFNO₄
MW: 278.03 g/mol
MDL Number: MFCD21606583
CAS Number: 1220886-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR