2-Bromo-5-(trifluoromethyl)pyridin-3-amine 98%

Supplier: Ambeed
Warning

A144056-100MG A144056-1G A144056-250MG A144056-5G
77027-746EA 48.81 USD
77027-746 77027-748 77027-750 77027-752
2-Bromo-5-(trifluoromethyl)pyridin-3-amine 98%
2-Bromo-5-(trifluoromethyl)pyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₄BrF₃N₂
MW: 241.009 g/mol
MDL Number: MFCD14702313
CAS Number: 1211515-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR