N-Boc-2-amino-5-bromo[1,3,4]thiadiazole 96%

Supplier: Ambeed
Warning

Synonyms: N-Boc-2-Amino-5-bromo[1,3,4]thiadiazole

A143981-100MG A143981-1G A143981-250MG A143981-25G A143981-5G
77027-736EA 20.16 USD
77027-736 77027-738 77027-740 77027-742 77027-744
N-Boc-2-amino-5-bromo[1,3,4]thiadiazole 96%
N-Boc-2-amino-5-bromo[1,3,4]thiadiazole

Precaución: for Research Use Only

Formula: C₇H₁₀BrN₃O₂S
MW: 280.1422 g/mol
MDL Number: MFCD11848382
CAS Number: 1048358-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR