2,6-Dibromo-5-chloronicotinic acid 95%

Supplier: Ambeed
Warning

A143838-100MG A143838-250MG
76984-314EA 13 USD
76984-314 76984-316
2,6-Dibromo-5-chloronicotinic acid 95%
2,6-Dibromo-5-chloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₂Br₂ClNO₂
MDL Number: MFCD26743442
CAS Number: 55304-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR