(S)-2-((4-Chlorophenyl)(piperidin-4-yloxy)methyl)pyridine 98%

Supplier: Ambeed
Warning

A143772-1G A143772-5G
77027-728EA 18.04 USD
77027-728 77027-730
(S)-2-((4-Chlorophenyl)(piperidin-4-yloxy)methyl)pyridine 98%
(S)-2-((4-Chlorophenyl)(piperidin-4-yloxy)methyl)pyridine

Precaución: for Research Use Only

Formula: C₁₇H₁₉ClN₂O
MW: 302.79 g/mol
MDL Number: MFCD09955359
CAS Number: 201594-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR