6-Bromo-5-nitroquinoline 97%

Supplier: Ambeed
Warning

A143618-100MG A143618-1G A143618-250MG A143618-5G
77027-716EA 28.65 USD
77027-716 77027-718 77027-720 77027-722
6-Bromo-5-nitroquinoline 97%
6-Bromo-5-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrN₂O₂
MW: 253.0500 g/mol
MDL Number: MFCD00837754
CAS Number: 98203-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR