5,6-Diamino-1,3-dipropyluracil hydrochloride 96%
Supplier: Ambeed
|
Warning
|
A143610-10G
A143610-1G
A143610-25G
A143610-5G
A143610-250MG
77027-708EA
100
USD
77027-708
77027-710
77027-712
77027-714
77667-068
5,6-Diamino-1,3-dipropyluracil hydrochloride 96%
5,6-Diamino-1,3-dipropyluracil hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₁₈N₄O₂·HCl |
MDL Number:
MFCD06797515 CAS Number: 324002-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...