Amiodarone hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Amiodarone HCl, (2-Butylbenzofuran-3-yl)(4-(2-(diethylamino)ethoxy)-3,5-diiodophenyl)methanone hydrochloride, 2-Butyl-3-benzofuranyl 4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl ketone hydrochloride)

A143592-1G A143592-1MG A143592-250MG A143592-25G A143592-5G
76996-978EA 24 USD
76996-978 76996-980 76996-982 76996-984 77600-072
Amiodarone hydrochloride 97%
Amiodarone hydrochloride

Precaución: for Research Use Only

Formula: C₂₅H₂₉I₂NO₃
MDL Number: MFCD00069204
CAS Number: 19774-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR