7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone 97%

Supplier: Ambeed
Warning

Synonyms: 7-(4-Bromobutoxy)-3,4-dihydroquinolin-2(1H)-one, 7-(4-Bromobutoxy)-1,2,3,4-tetrahydro-2-oxoquinoline

A143576-100G A143576-10G A143576-1G A143576-250MG A143576-25G A143576-5G
76996-966EA 290.69 USD
76996-966 76996-968 76996-970 76996-972 76996-974 76996-976
7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone 97%
7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrNO₂
MW: 298.18 g/mol
MDL Number: MFCD06658540
CAS Number: 129722-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR