p-Anisic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxybenzoic acid, 4-Anisic acid

A143500-1000G A143500-100G A143500-25G A143500-500G
77027-698EA 53.05 USD
77027-698 77027-700 77027-702 77027-704
p-Anisic acid 98%
p-Anisic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00002542
CAS Number: 100-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR