Ethyl 3-iodo-1H-pyrazole-4-carboxylate 98%

Supplier: Ambeed
Warning

A143473-1G A143473-250MG A143473-5G
77027-692EA 16 USD
77027-692 77027-694 77027-696
Ethyl 3-iodo-1H-pyrazole-4-carboxylate 98%
Ethyl 3-iodo-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇IN₂O₂
MDL Number: MFCD18072591
CAS Number: 827316-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR