1-Oxa-4,9-diazaspiro[5.5]undecane, N9-BOC protected 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 1-oxa-4,9-diazaspiro[5.5]undecane-9-carboxylate

A143327-100MG A143327-1G A143327-250MG A143327-5G
77027-680EA 21.22 USD
77027-680 77027-682 77027-684 77027-686
1-Oxa-4,9-diazaspiro[5.5]undecane, N9-BOC protected 97%
1-Oxa-4,9-diazaspiro[5.5]undecane, N9-BOC protected

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₃
MW: 256.341 g/mol
MDL Number: MFCD11227065
CAS Number: 930785-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR