2-Amino-5-bromo-4-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4-(trifluoromethyl)pyridin-2-amine

A143318-10G A143318-1G A143318-250MG A143318-25G A143318-5G
76996-944EA 67 USD
76996-944 76996-946 76996-948 76996-950 76996-952
2-Amino-5-bromo-4-(trifluoromethyl)pyridine 97%
2-Amino-5-bromo-4-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrF₃N₂
MDL Number: MFCD09864896
CAS Number: 944401-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR