Methyl 6-bromo-5-nitronicotinate 95%

Supplier: Ambeed
Warning

A143209-100MG A143209-1G A143209-250MG A143209-5G
76984-284EA 99 USD
76984-284 76984-286 76984-288 76984-290
Methyl 6-bromo-5-nitronicotinate 95%
Methyl 6-bromo-5-nitronicotinate

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O₄
MDL Number: MFCD18257551
CAS Number: 1211519-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR