ɑ-(4-Chlorophenyl)-2-pyridinemethanol 97%

Supplier: Ambeed
Warning

A143082-100G A143082-10G A143082-25G A143082-5G
77027-668EA 169 USD
77027-668 77027-670 77027-672 77027-674
ɑ-(4-Chlorophenyl)-2-pyridinemethanol 97%
alpha-(4-Chlorophenyl)-2-pyridinemethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO
MDL Number: MFCD00023493
CAS Number: 27652-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR