Sunitinib malate 98%

Supplier: Ambeed
Danger

Synonyms: (Z)-N-(2-(Diethylamino)ethyl)-5-((5-fluoro-2-oxoindolin-3-ylidene)methyl)-2,4-dimethyl-1H-pyrrole-3-carboxamide (S)-2-hydroxysuccinate, SU-11248

A142950-100MG A142950-1G A142950-250MG A142950-25G A142950-25MG A142950-5G
77027-656EA 6.37 USD
77027-656 77027-658 77027-660 77027-662 77027-664 77027-666
Sunitinib malate 98%
Sunitinib malate

Precaución: for Research Use Only

Formula: C₂₂H₂₇FN₄O₂·C₄H₆O₅
MW: 532.6 g/mol
MDL Number: MFCD08282795
CAS Number: 341031-54-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR