5-Fluoro-4-methyl-2-nitrophenol 98%

Supplier: Ambeed
Danger

A142937-250MG A142937-5G
77010-458EA 110.34 USD
77010-458 77010-460
5-Fluoro-4-methyl-2-nitrophenol 98%
5-Fluoro-4-methyl-2-nitrophenol

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
MW: 171.126 g/mol
MDL Number: MFCD16658619
CAS Number: 83341-28-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR