5-Amino-8-fluoroquinoline 98%

Supplier: Ambeed
Warning

Synonyms: 8-Fluoroquinolin-5-amine

A142892-1G A142892-250MG A142892-100MG
77027-652EA 326.76 USD
77027-652 77027-654 77671-038
5-Amino-8-fluoroquinoline 98%
5-Amino-8-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₇FN₂
MW: 162.1637 g/mol
MDL Number: MFCD09042730
CAS Number: 175229-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR