3-Amino-5-fluoropyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoropyridin-3-amine

A142880-100G A142880-1G A142880-5G
76996-932EA 257.8 USD
76996-932 76996-934 76996-936
3-Amino-5-fluoropyridine 97%
3-Amino-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₅FN₂
MW: 112.11 g/mol
MDL Number: MFCD00234073
CAS Number: 210169-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR