7-Bromo-1-methylquinolin-2(1H)-one 95%

Supplier: Ambeed
Warning

A142857-1G A142857-250MG
77010-452EA 222 USD
77010-452 77010-454
7-Bromo-1-methylquinolin-2(1H)-one 95%
7-Bromo-1-methylquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MDL Number: MFCD12913830
CAS Number: 1187933-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR