2-Aminopyrimidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyrimidin-2-ylamine, Pyrimidin-2-amine
A142818-1000G
A142818-100G
A142818-10G
A142818-25G
A142818-500G
77027-640EA
162.32
USD
77027-640
77027-642
77027-644
77027-646
77027-648
2-Aminopyrimidine 98%
2-Aminopyrimidine
Precaución: for Research Use Only
Formula:
C₄H₅N₃ MW: 95.1 g/mol |
MDL Number:
MFCD00006089 CAS Number: 109-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...