3-Oxo-3,4-dihydropyrazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A142795-100G A142795-10G A142795-1G A142795-25G A142795-5G
77010-436EA 52 USD
77010-436 77010-438 77010-440 77010-442 77010-444
3-Oxo-3,4-dihydropyrazine-2-carboxylic acid 97%
3-Oxo-3,4-dihydropyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C5H4N2O3
MDL Number: MFCD00235136
CAS Number: 20737-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR