2-Imidazol-1-ylethylamine dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Aminoethyl)-1H-imidazole dihydrochloride, 2-(1H-Imidazol-1-yl)ethanamine dihydrochloride

A142722-100MG A142722-1G A142722-250MG
76984-274EA 21 USD
76984-274 76984-276 76984-278
2-Imidazol-1-ylethylamine dihydrochloride 98%
2-Imidazol-1-ylethylamine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁Cl₂N₃
MDL Number: MFCD06798214
CAS Number: 93668-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR