(4-Methoxyphenoxy)acetic acid 97%

Supplier: Ambeed
Warning

A142716-1G A142716-250MG A142716-25G A142716-5G
76996-922EA 40.32 USD
76996-922 76996-924 76996-926 76996-928 76996-930
(4-Methoxyphenoxy)acetic acid 97%
(4-Methoxyphenoxy)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄(OCH₃))OCH₂COOH
MDL Number: MFCD00014360
CAS Number: 1877-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR