Methyl-2-((tert-butoxycarbonyl)amino)-2-(piperidin-4-yl)acetate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-((tert-butoxycarbonyl)amino)-2-(piperidin-4-yl)acetate

A142650-100MG A142650-250MG A142650-1G
77027-636EA 66 USD
77027-636 77027-638 77592-694
Methyl-2-((tert-butoxycarbonyl)amino)-2-(piperidin-4-yl)acetate 95%
Methyl-2-((tert-butoxycarbonyl)amino)-2-(piperidin-4-yl)acetate

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₄
MDL Number: MFCD04115491
CAS Number: 219832-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR