9-([1,1'-Biphenyl]-4-yl)-9H,9'H-3,3'-bicarbazole 98%

Supplier: Ambeed
Warning

A142637-1G A142637-250MG A142637-5G
77010-422EA 35.01 USD
77010-422 77010-424 77010-426
9-([1,1'-Biphenyl]-4-yl)-9H,9'H-3,3'-bicarbazole 98%
9-([1,1'-Biphenyl]-4-yl)-9H,9'H-3,3'-bicarbazole

Precaución: for Research Use Only

Formula: C₃₆H₂₄N₂
MW: 484.58 g/mol
MDL Number: MFCD30536715
CAS Number: 1346669-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR