9-(4-Biphenylyl)-3-bromocarbazole 97%

Supplier: Ambeed
Warning

Synonyms: 9-([1,1'-Biphenyl]-4-yl)-3-bromo-9H-carbazole

A142586-100G A142586-1G A142586-25G A142586-5G
76984-256EA 124 USD
76984-256 76984-258 76984-260 76984-262
9-(4-Biphenylyl)-3-bromocarbazole 97%
9-(4-Biphenylyl)-3-bromocarbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₆BrN
MDL Number: MFCD16038226
CAS Number: 894791-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR