(E)-3-(2-Cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl)acrylonitrile 97%

Supplier: Ambeed
Warning

A142388-100MG A142388-250MG A142388-5G
76984-248EA 6.37 USD
76984-248 76984-250 76984-252
(E)-3-(2-Cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl)acrylonitrile 97%
(E)-3-(2-Cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl)acrylonitrile

Precaución: for Research Use Only

Formula: C₂₁H₁₅FN₂
MW: 314.36 g/mol
MDL Number: MFCD11042688
CAS Number: 256431-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR