1-(5-Bromothien-2-yl)ethanamine 96%

Supplier: Ambeed
Warning

A142340-1G A142340-250MG A142340-5G
77027-620EA 103 USD
77027-620 77027-622 77027-624
1-(5-Bromothien-2-yl)ethanamine 96%
1-(5-Bromothien-2-yl)ethanamine

Precaución: for Research Use Only

Formula: C₆H₈BrNS
MDL Number: MFCD05215403
CAS Number: 129605-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR