2,5-Dimethoxypyrimidin-4(3H)-one 98%

Supplier: Ambeed
Warning

A142315-10G A142315-25G
76984-244EA 9.55 USD
76984-244 76984-246
2,5-Dimethoxypyrimidin-4(3H)-one 98%
2,5-Dimethoxypyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₆H₈N₂O₃
MW: 156.14 g/mol
MDL Number: MFCD20480733
CAS Number: 370103-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR