4-Bromo-2-iodo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A142022-100MG A142022-250MG
76984-228EA 45 USD
76984-228 76984-230
4-Bromo-2-iodo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine 95%
4-Bromo-2-iodo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₃H₈BrIN₂O₂S
MDL Number: MFCD08443802
CAS Number: 889939-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR