3-Chloro-6-methylbenzo[b]thiophene-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A141733-1G
A141733-250MG
A141733-5G
76996-888EA
30
USD
76996-888
76996-890
76996-892
3-Chloro-6-methylbenzo[b]thiophene-2-carboxylic acid 95%
3-Chloro-6-methylbenzo[b]thiophene-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₇ClO₂S |
MDL Number:
MFCD00687576 CAS Number: 34576-96-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...