4,6-Dichloro-5-ethyl-2-(methylsulfonyl)pyrimidine 95%

Supplier: Ambeed
Warning

A141656-1G A141656-250MG
76984-220EA 369.2 USD
76984-220 76984-222
4,6-Dichloro-5-ethyl-2-(methylsulfonyl)pyrimidine 95%
4,6-Dichloro-5-ethyl-2-(methylsulfonyl)pyrimidine

Precaución: for Research Use Only

Formula: C₇H₈Cl₂N₂O₂S
MW: 255.12 g/mol
MDL Number: MFCD11846228
CAS Number: 1263314-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR