Methyl-4-bromo-3-(trifluoromethyl)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-3-(trifluoromethyl)benzoate, 4-Bromo-3-(trifluoromethyl)benzoic acid methyl ester

A141504-100G A141504-10G A141504-1G A141504-250MG A141504-25G A141504-5G
77027-564EA 434.97 USD
77027-564 77027-566 77027-568 77027-570 77027-572 77027-574
Methyl-4-bromo-3-(trifluoromethyl)benzoate 98%
Methyl 4-bromo-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₂
MW: 283.0420 g/mol
MDL Number: MFCD10566409
CAS Number: 107317-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR