Methyl 4-amino-2,5-difluorobenzoate 95%

Supplier: Ambeed
Warning

A141391-100MG A141391-10G A141391-1G A141391-250MG A141391-25G A141391-5G
77010-356EA 5 USD
77010-356 77010-358 77010-360 77010-362 77010-364 77010-366
Methyl 4-amino-2,5-difluorobenzoate 95%
Methyl 4-amino-2,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇F₂NO₂
MDL Number: MFCD20725922
CAS Number: 952285-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR