4-Bromo-1H-pyrrole-2-carboxamide 97%

Supplier: Ambeed
Warning

A141388-100MG A141388-1G A141388-250MG
76984-206EA 40.32 USD
76984-206 76984-208 76984-210
4-Bromo-1H-pyrrole-2-carboxamide 97%
4-Bromo-1H-pyrrole-2-carboxamide

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
MW: 189.01 g/mol
MDL Number: MFCD11504939
CAS Number: 196106-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR