2-Amino-5-nitrobenzophenone 98%

Supplier: Ambeed
Warning

A141234-100G A141234-10G A141234-25G A141234-500G A141234-5G
76996-876EA 51 USD
76996-876 76996-878 76996-880 76996-882 77671-016
2-Amino-5-nitrobenzophenone 98%
2-Amino-5-nitrobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂O₃
MDL Number: MFCD00007364
CAS Number: 1775-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR