Parecoxib Sodium 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Dynastat, SC-69124A, Sodium ((4-(5-methyl-3-phenylisoxazol-4-yl)phenyl)sulphonyl)(propionyl)amide
A141185-100MG
A141185-1MG
A141185-25MG
A141185-50MG
A141185-5MG
A141185-1G
A141185-250MG
A141185-5G
77027-530EA
144.29
USD
77027-530
77027-532
77027-534
77027-536
77027-538
77667-038
77667-040
77667-042
Parecoxib Sodium 98%
Parecoxib sodium
Precaución: for Research Use Only
Formula:
C₁₉H₁₇N₂NaO₄S MW: 392.4 g/mol |
MDL Number:
MFCD04972608 CAS Number: 198470-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...