1-Benzyl 2-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Cbz-D-cis-Hyp-OMe

A141146-1G A141146-250MG A141146-5G A141146-25G
77027-526EA 37.14 USD
77027-526 77027-528 77592-674 77600-050
1-Benzyl 2-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate 95%
1-Benzyl 2-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₅
MW: 279.28 g/mol
MDL Number: MFCD09954933
CAS Number: 155075-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR