2-Bromo-4-chloro-3-fluoropyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-chloro-3-fluoro-pyridine

A141043-100MG A141043-1G A141043-250MG A141043-5G
76984-182EA 60.48 USD
76984-182 76984-184 76984-186 76984-188
2-Bromo-4-chloro-3-fluoropyridine 95%
2-Bromo-4-chloro-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
MW: 210.43 g/mol
MDL Number: MFCD18257572
CAS Number: 1155847-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR