2-(4-(Trifluoromethoxy)phenyl)ethanamine 98%

Supplier: Ambeed
Danger

A141040-25G A141040-5G
76996-870EA 185.66 USD
76996-870 77671-014
2-(4-(Trifluoromethoxy)phenyl)ethanamine 98%
2-(4-(Trifluoromethoxy)phenyl)ethanamine

Precaución: for Research Use Only

Formula: C₉H₁₀F₃NO
MW: 205.18 g/mol
MDL Number: MFCD06212783
CAS Number: 170015-99-3
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR