5-(4-Ethylbenzylidene)-2-thioxothiazolidin-4-one 98+%

Supplier: Ambeed
Warning

Synonyms: 10058-F4

A140998-10MG A140998-1MG A140998-5MG
77027-510EA 11.67 USD
77027-510 77027-512 77027-514
5-(4-Ethylbenzylidene)-2-thioxothiazolidin-4-one 98+%
5-(4-Ethylbenzylidene)-2-thioxothiazolidin-4-one

Precaución: for Research Use Only

Formula: C₁₂H₁₁NOS₂
MW: 249.35 g/mol
MDL Number: MFCD04969046
CAS Number: 403811-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR