Naphthalene-1,8-diyl dibenzoate 98%

Supplier: Ambeed
Warning

A140847-100MG A140847-250MG
77010-338EA 21.22 USD
77010-338 77010-340
Naphthalene-1,8-diyl dibenzoate 98%
Naphthalene-1,8-diyl dibenzoate

Precaución: for Research Use Only

Formula: C₂₄H₁₆O₄
MW: 368.38 g/mol
MDL Number: MFCD01001009
CAS Number: 331711-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR