Benzyloxyacetic acid 97%

Supplier: Ambeed
Warning

A140838-100G A140838-10G A140838-1G A140838-25G A140838-5G A140838-500G
76996-860EA 39 USD
76996-860 76996-862 76996-864 76996-866 76996-868 77881-100
Benzyloxyacetic acid 97%
Benzyloxyacetic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD00274211
CAS Number: 30379-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR