2,3-Dimethyl-5-nitroaniline 95%

Supplier: Ambeed
Warning

A140527-1G A140527-250MG
76984-156EA 65 USD
76984-156 76984-158
2,3-Dimethyl-5-nitroaniline 95%
2,3-Dimethyl-5-nitroaniline

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD05664520
CAS Number: 109508-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR